Please login to your online account to display your discounted pricing

2-Nitro-4-(trifluoromethyl)phenol, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenol
CAS 400-99-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H4F3NO3
Linear Formula O2NC6H3(CF3)OH
Formula Weight 207.11
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC218880010

  SDS
thermo fisher scientific
AC218880010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC218880050

  SDS
thermo fisher scientific
AC218880050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5%
Formula Weight 207.11
Quantity 1g
Color Yellow to orange
Density 1.47g/mL
CAS Max % 100.0
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenol
Assay Percent Range 99%
Molecular Formula C7H4F3NO3
Linear Formula O2NC6H3(CF3)OH
CAS 400-99-7
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.