Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitro-4-(trifluoromethyl)benzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzonitrile
CAS 778-94-9
Molecular Formula C8H3F3N2O2
Formula Weight 216.12
MDL Number MFCD00014684
UN Number UN3439
Beilstein 1882259
Synonym 4-Cyano-3-nitrobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1055706

 
alfa aesar™
L1055706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1055714

 
alfa aesar™
L1055714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.12
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzonitrile
Assay 98%
Molecular Formula C8H3F3N2O2
CAS 778-94-9
Beilstein 1882259
Synonym 4-Cyano-3-nitrobenzotrifluoride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Quantity 5g
MDL Number MFCD00014684
UN Number UN3439
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Flash Point >110°C (230°F)
Boiling Point 156° to 158°C (18mmHg)
Melting Point 40° to 44°C
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.