missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitro-4-(trifluoromethyl)benzoic acid, 98%

Manufacturer:  Alfa Aesar™ 4378702

 View more versions of this product

Catalog No. AA4378702



Specifications

Specifications

2-Nitro-4- (trifluoromethyl)benzoic acid
320-94-5
MFCD00007142
MYSAXQPTXWKDPQ-UHFFFAOYSA-N
235.118
235.12
98%
0.5g
C8H4F3NO4
2-nitro-4-trifluoromethyl benzoic acid
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
642096
Powder

Chemical Identifiers

320-94-5
235.118
MYSAXQPTXWKDPQ-UHFFFAOYSA-N
642096
C8H4F3NO4
MFCD00007142
2-nitro-4-trifluoromethyl benzoic acid
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents