Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitro-4-(trifluoromethyl)anisole, 98%

Chemical Identifiers

Chemical Name or Material 2-Nitro-4- (trifluoromethyl)anisole
CAS 394-25-2
Molecular Formula C8H6F3NO3
Formula Weight 221.14
MDL Number MFCD00007099
UN Number UN1759
Beilstein 2127954
Synonym 4-Methoxy-3-nitrobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1184714

 
alfa aesar™
A1184714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1184722

 
alfa aesar™
A1184722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.14
Chemical Name or Material 2-Nitro-4- (trifluoromethyl)anisole
Assay 98%
Molecular Formula C8H6F3NO3
MDL Number MFCD00007099
Boiling Point 96° to 98°C (0.6mmHg)
CAS 394-25-2
UN Number UN1759
Beilstein 2127954
Melting Point 44° to 49°C
Synonym 4-Methoxy-3-nitrobenzotrifluoride
Quantity 25g
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.