missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Nitro-1-propanol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04384306
Description
2-Nitro-1-propanol is used in improved synthesis of 2,2,2-trinitroethyl ethers, inhibits methane production during in vitro ruminal fermentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Nitro-1-propanol | |
1.185 g/mL | |
100°C (212°F) | |
1.439 | |
5 g | |
Slightly soluble in water. | |
CC(CO)[N+]([O-])=O | |
105.09 | |
105.09 | |
Liquid |
2902-96-7 | |
73°C (1 mmHg) | |
C3H7NO3 | |
MFCD00007398 | |
2-nitro-1-propanol, 2-nitropropanol, 1-propanol, 2-nitro, 2-nitropropanol-1, ccris 5045, 2-nitro-propan-1-ol, acmc-20anp9, 1-hydroxy-2-nitropropane, 1-propanol, 2-nitro-8ci | |
PCNWBUOSTLGPMI-UHFFFAOYNA-N | |
2-nitropropan-1-ol | |
92187 | |
98% |
Chemical Identifiers
2902-96-7 | |
105.09 | |
PCNWBUOSTLGPMI-UHFFFAOYNA-N | |
92187 | |
CC(CO)[N+]([O-])=O |
C3H7NO3 | |
MFCD00007398 | |
2-nitro-1-propanol, 2-nitropropanol, 1-propanol, 2-nitro, 2-nitropropanol-1, ccris 5045, 2-nitro-propan-1-ol, acmc-20anp9, 1-hydroxy-2-nitropropane, 1-propanol, 2-nitro-8ci | |
2-nitropropan-1-ol |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only