missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Nitro-1-propanol, 98%

Manufacturer:  Alfa Aesar™ 4384314

 View more versions of this product

Catalog No. AA4384314



Specifications

Specifications

2-Nitro-1-propanol
100°C (212°F)
73°C (1mmHg)
2902-96-7
MFCD00007398
PCNWBUOSTLGPMI-UHFFFAOYSA-N
2-nitropropan-1-ol
92187
Liquid
1.185g/mL
1.4390
25g
C3H7NO3
1-hydroxy-2-nitropropane
CC(CO)[N+](=O)[O-]
105.093
105.09
98%

Chemical Identifiers

2902-96-7
105.093
PCNWBUOSTLGPMI-UHFFFAOYSA-N
92187
CC(CO)[N+](=O)[O-]
C3H7NO3
MFCD00007398
1-hydroxy-2-nitropropane
2-nitropropan-1-ol
Documents