missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthylmagnesium bromide, 0.25M in MeTHF

$203.19 - $203.19

Chemical Identifiers

CAS 21473-01-8
Molecular Formula C10H7BrMg
Formula Weight 231.37
MDL Number MFCD01319910
UN Number UN2924
Chemical Name or Material 2-Naphthylmagnesium bromide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH51160AD SDS Alfa Aesar™
H51160AD
50mL Each for $203.19
AAH51160AE SDS Alfa Aesar™
H51160AE
100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

21473-01-8
231.37
UN2924
C10H7BrMg
MFCD01319910
2-Naphthylmagnesium bromide

Specifications

21473-01-8
231.37
MFCD01319910
50mL
C10H7BrMg
0.25M in MeTHF
UN2924
2-Naphthylmagnesium bromide

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.