Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthylacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 2-Naphthylacetonitrile
CAS 7498-57-9
Molecular Formula C12H9N
Formula Weight 167.21
MDL Number MFCD00004122
UN Number UN3439
Beilstein 1862879
Synonym 2-Naphthaleneacetonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0938406

  SDS
alfa aesar™
L0938406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0938414

  SDS
alfa aesar™
L0938414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.21
Molecular Formula C12H9N
Chemical Name or Material 2-Naphthylacetonitrile
Synonym 2-Naphthaleneacetonitrile
CAS 7498-57-9
MDL Number MFCD00004122
Health Hazard 3 GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Assay 97%
UN Number UN3439
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Quantity 5g
Boiling Point 303°C
Melting Point 82° to 84°C
Beilstein 1862879

Applications
2-Naphthylacetonitrile was used as starting reagent in the synthesis of 2-(1-cyano-1-(2-naphthyl))methylidene-3-phenyl-thiazolidine-4,5-dione.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.