Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthylacetic acid, 99%

$102.77 - $409.84

Chemical Identifiers

Chemical Name or Material 2-Naphthylacetic acid
CAS 581-96-4
Molecular Formula C12H10O2
Formula Weight 186.21
MDL Number MFCD00004126
Beilstein 973396
Synonym 2-Naphthaleneacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1352809

  SDS
alfa aesar™
A1352809
10g Each for $102.77

AAA1352818

  SDS
alfa aesar™
A1352818
50g Each for $409.84
Description & Specifications

Specifications

Formula Weight 186.21
Chemical Name or Material 2-Naphthylacetic acid
Assay 99%
Molecular Formula C12H10O2
MDL Number MFCD00004126
CAS 581-96-4
Beilstein 973396
Synonym 2-Naphthaleneacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number QJ0876000
Melting Point 141° to 143°C
Quantity 10g

Applications
2-Naphthylacetic acid is a synthetic plant hormone in the auxin family and is an ingredient in many commercial plant rooting horticultural products, it is a rooting agent and used for the vegetative propagation of plants from stem and leaf cutting. It is also used for plant tissue culture.

Solubility
Soluble in acetone (50 mg/ml), and methanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.