missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthyl trifluoromethanesulfonate, 97%

Manufacturer:  Alfa Aesar™ H5541703

 View more versions of this product

Catalog No. AAH5541703



Specifications

Specifications

2-Naphthyl trifluoromethanesulfonate
Orange to White
1g
C11H7F3O3S
UN3261
MDWRQYBWVTXIIJ-UHFFFAOYSA-N
naphthalen-2-yl trifluoromethanesulfonate
4546341
Solid
113°C (235°F)
30°C to 32°C
3857-83-8
MFCD00192341
2-naphthyl triflate
C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)C(F)(F)F
276.229
276.23
97%

Chemical Identifiers

3857-83-8
276.229
MDWRQYBWVTXIIJ-UHFFFAOYSA-N
4546341
C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)C(F)(F)F
C11H7F3O3S
MFCD00192341
2-naphthyl triflate
naphthalen-2-yl trifluoromethanesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents