Learn More
2-Naphthyl trifluoromethanesulfonate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5541706
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Naphthyl trifluoromethanesulfonate | |
30°C to 32°C | |
113°C (235°F) | |
MFCD00192341 | |
UN3261 | |
MDWRQYBWVTXIIJ-UHFFFAOYSA-N | |
naphthalen-2-yl trifluoromethanesulfonate | |
4546341 | |
97% |
3857-83-8 | |
Orange to White | |
C11H7F3O3S | |
5 g | |
2-naphthyl trifluoromethanesulfonate, 2-naphthyl triflate, 2-naphthyltrifluoromethanesulfonate, 2-naphthyltriflate, acmc-1aigh, trifluoromethanesulfonic acid 2-naphthyl ester, methanesulfonic acid, 1,1,1-trifluoro-, 2-naphthalenyl ester, methanesulfonic acid,1,1,1-trifluoro-, 2-naphthalenyl ester | |
C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)C(F)(F)F | |
276.229 | |
276.23 | |
Solid |
Chemical Identifiers
3857-83-8 | |
276.229 | |
MDWRQYBWVTXIIJ-UHFFFAOYSA-N | |
4546341 | |
C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)C(F)(F)F |
C11H7F3O3S | |
MFCD00192341 | |
2-naphthyl trifluoromethanesulfonate, 2-naphthyl triflate, 2-naphthyltrifluoromethanesulfonate, 2-naphthyltriflate, acmc-1aigh, trifluoromethanesulfonic acid 2-naphthyl ester, methanesulfonic acid, 1,1,1-trifluoro-, 2-naphthalenyl ester, methanesulfonic acid,1,1,1-trifluoro-, 2-naphthalenyl ester | |
naphthalen-2-yl trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures