Please login to your online account to display your discounted pricing

2-Naphthyl sulfate, potassium salt, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  230250010

 View more versions of this product

Catalog No. AC230250010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 262.32
Quantity 1g
Color White to beige
EINECS Number 244-375-0
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00004059
CAS Min % 98.5
Molecular Formula C10 H7 K O 4S
Linear Formula C10H7OSO3K
CAS 21409-32-5
Chemical Name or Material 2-Naphthyl sulfate, potassium salt
Name Note 99%
Assay Percent Range 99%

Specifications

Chemical Name or Material 2-Naphthyl sulfate, potassium salt
Name Note 99%
CAS 21409-32-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10 H7 K O 4S
Linear Formula C10H7OSO3K
Formula Weight 262.32
MDL Number MFCD00004059