missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Naphthyl sulfate, potassium salt, 99%, ACROS Organics™

Chemical Identifiers

CAS 21409-32-5
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C10 H7 K O 4S
Linear Formula C10H7OSO3K
Formula Weight 262.32
MDL Number MFCD00004059
Chemical Name or Material 2-Naphthyl sulfate, potassium salt, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC230250010 SDS Thermo Fisher Scientific
230250010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

21409-32-5
100.0
C10 H7 K O 4S
262.32
2-Naphthyl sulfate, potassium salt, 99%
98.5
99%
C10H7OSO3K
MFCD00004059
≥98.5%

Specifications

21409-32-5
100.0
Glass bottle
99%
C10H7OSO3K
MFCD00004059
1g
≥98.5%
98.5
Authentic
Fine Crystalline Powder
C10 H7 K O 4S
262.32
White to beige
2-Naphthyl sulfate, potassium salt, 99%

Safety and Handling

EINECSNumber : 244-375-0

TSCA : TSCA