missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthyl sulfate potassium salt, 98%

$60.91 - $177.99

Chemical Identifiers

CAS 1733-89-7
Molecular Formula C10H8KO4S
Molecular Weight (g/mol) 263.328
MDL Number MFCD00004059
InChI Key KPQVGLIXPMOHMI-UHFFFAOYSA-N
Synonym 2-naphthyl hydrogen sulfate
PubChem CID 24186395
IUPAC Name naphthalen-2-yl hydrogen sulfate;potassium
SMILES C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)O.[K]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA44061MD View Documents Alfa Aesar™
44061MD
250mg Each for $60.91
 
AA4406103 View Documents Alfa Aesar™
4406103
1g Each for $177.99
 
Specifications

Chemical Identifiers

1733-89-7
263.328
KPQVGLIXPMOHMI-UHFFFAOYSA-N
24186395
C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)O.[K]
C10H8KO4S
MFCD00004059
2-naphthyl hydrogen sulfate
naphthalen-2-yl hydrogen sulfate;potassium

Specifications

>300°C
1733-89-7
MFCD00004059
KPQVGLIXPMOHMI-UHFFFAOYSA-N
naphthalen-2-yl hydrogen sulfate;potassium
24186395
Powder
2-Naphthyl sulfate potassium salt
250mg
C10H8KO4S
2-naphthyl hydrogen sulfate
C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)O.[K]
263.328
262.33
98%
Safety and Handling

Safety and Handling

EINECSNumber : 217-072-6

TSCA : Yes

Recommended Storage : Ambient temperatures