missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthyl sulfate potassium salt, 98%

Manufacturer:  Alfa Aesar™ 44061MD

 View more versions of this product

Catalog No. AA44061MD



Specifications

Specifications

2-Naphthyl sulfate potassium salt
250mg
C10H8KO4S
2-naphthyl hydrogen sulfate
C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)O.[K]
263.328
262.33
98%
>300°C
1733-89-7
MFCD00004059
KPQVGLIXPMOHMI-UHFFFAOYSA-N
naphthalen-2-yl hydrogen sulfate;potassium
24186395
Powder

Chemical Identifiers

1733-89-7
263.328
KPQVGLIXPMOHMI-UHFFFAOYSA-N
24186395
C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)O.[K]
C10H8KO4S
MFCD00004059
2-naphthyl hydrogen sulfate
naphthalen-2-yl hydrogen sulfate;potassium
Safety and Handling

Safety and Handling

EINECSNumber : 217-072-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents