Please login to your online account to display your discounted pricing

2-Naphthyl Phenyl Ketone 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  415420250

 View more versions of this product

Catalog No. AC415420250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 232.28
Physical Form Powder
Quantity 25g
Color White to pale yellow
Melting Point 76° to 82°C
TSCA TSCA
EINECS Number 211-410-6
CAS Max % 100.0
Health Hazard 1 PRODUCT NEEDS INERT ATMOSPHERE
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C17H12O
MDL Number MFCD00004106
Chemical Name or Material 2-Naphthyl phenyl ketone
Name Note 98%
CAS 644-13-3
Assay Percent Range 98%

Specifications

Chemical Name or Material 2-Naphthyl phenyl ketone
Name Note 98%
CAS 644-13-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C17H12O
Formula Weight 232.28
MDL Number MFCD00004106
Accessories

PRODUCT NEEDS INERT ATMOSPHERE

SDS & Safety Documents