Please login to your online account to display your discounted pricing

2-Naphthyl Phenyl Ketone 98%, ACROS Organics™

$213.42 - $710.29

Chemical Identifiers

Chemical Name or Material 2-Naphthyl phenyl ketone
Name Note 98%
CAS 644-13-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C17H12O
Formula Weight 232.28
MDL Number MFCD00004106
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC415420050

  SDS
thermo fisher scientific
415420050
Glass bottle 5g Each for $213.42

AC415420250

  SDS
thermo fisher scientific
415420250
Glass bottle 25g Each for $710.29
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 232.28
Physical Form Powder
Quantity 5g
Color White to pale yellow
TSCA TSCA
Melting Point 76° to 82°C
EINECS Number 211-410-6
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 1 PRODUCT NEEDS INERT ATMOSPHERE
Assay Percent Range 98%
Name Note 98%
Chemical Name or Material 2-Naphthyl phenyl ketone
Molecular Formula C17H12O
CAS 644-13-3
MDL Number MFCD00004106
Safety and Handling

PRODUCT NEEDS INERT ATMOSPHERE