Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2-Naphthyl isocyanate
CAS 2243-54-1
Molecular Formula C11H7NO
Formula Weight 169.18
MDL Number MFCD00014319
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5107603

  SDS
alfa aesar™
H5107603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.18
Quantity 1g
Assay 97%
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 1 Danger
Molecular Formula C11H7NO
Odor Pungent
Melting Point 53° to 56°C
UN Number UN2811
Chemical Name or Material 2-Naphthyl isocyanate
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H331-H334-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 2243-54-1
MDL Number MFCD00014319
Boiling Point 83°C (0.2mmHg)

Applications
2-Naphthyl isocyanate is used as pharmaceutical intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.

Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H334-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.