Please login to your online account to display your discounted pricing

2-Naphthyl benzoate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  330610250

 View more versions of this product

Catalog No. AC330610250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 248.28
Quantity 25g
Color Light brown
EINECS Number 202-247-1
TSCA TSCA
Melting Point 105° to 109°C
Merck Index 15, 6490
CAS Max % 100.0
Chemical Name or Material 2-Naphthyl benzoate
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C17H12O2
MDL Number MFCD00014320
CAS 93-44-7
CAS Min % 97.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Naphthyl benzoate
Name Note 98%
CAS 93-44-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C17H12O2
Formula Weight 248.28
MDL Number MFCD00014320