Please login to your online account to display your discounted pricing

2-Naphthyl benzoate, 98%, ACROS Organics™

$26.39 - $99.04

Chemical Identifiers

Chemical Name or Material 2-Naphthyl benzoate
Name Note 98%
CAS 93-44-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C17H12O2
Formula Weight 248.28
MDL Number MFCD00014320
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC330610050

  SDS
thermo fisher scientific
330610050
Glass bottle 5g Each for $26.39

AC330610250

  SDS
thermo fisher scientific
330610250
Glass bottle 25g Each for $99.04
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 248.28
Quantity 5g
Color Light brown
Merck Index 15, 6490
EINECS Number 202-247-1
TSCA TSCA
Melting Point 105° to 109°C
CAS Max % 100.0
Chemical Name or Material 2-Naphthyl benzoate
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C17H12O2
MDL Number MFCD00014320
CAS 93-44-7
CAS Min % 97.5
Infrared Spectrum Authentic