Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthyl benzoate, 98+%

$98.66 - $98.66

Chemical Identifiers

Chemical Name or Material 2-Naphthyl benzoate
CAS 93-44-7
Molecular Formula C17H12O2
Formula Weight 248.28
MDL Number MFCD00014320
Beilstein 2052424
Merck Index 14,6408
Synonym Benzoic acid 2-naphthyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1293514

  SDS
alfa aesar™
A1293514
25g Each for $98.66
Description & Specifications

Specifications

Formula Weight 248.28
Chemical Name or Material 2-Naphthyl benzoate
TSCA TSCA
Assay ≥98%
Molecular Formula C17H12O2
Melting Point 105° to 109°C
MDL Number MFCD00014320
CAS 93-44-7
Beilstein 2052424
Quantity 25g
Merck Index 14,6408
Odor Odorless
Synonym Benzoic acid 2-naphthyl ester