missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthoxyacetic acid, 97%

Manufacturer:  Alfa Aesar™ A154120E

 View more versions of this product

Catalog No. AAA154120E



Description

Description

Applications
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

Solubility
Soluble in alcohol, ether, acetic acid and dimethyl sulfoxide. Slightly soluble in water.

Notes
Store in a cool place. Incompatible with oxidizing agents.
Specifications

Specifications

2-Naphthoxyacetic acid
2500g
C12H10O3
1074148
2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal
C1=CC=C2C=C(C=CC2=C1)OCC(=O)O
202.209
CHEBI:50397
97%
155°C to 157°C
120-23-0
MFCD00004066
14,6397
RZCJYMOBWVJQGV-UHFFFAOYSA-N
2-naphthalen-2-yloxyacetic acid
8422
202.21

Chemical Identifiers

120-23-0
202.209
RZCJYMOBWVJQGV-UHFFFAOYSA-N
8422
2-naphthalen-2-yloxyacetic acid
C12H10O3
MFCD00004066
2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal
CHEBI:50397
C1=CC=C2C=C(C=CC2=C1)OCC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AI9659000

TSCA : TSCA

Documents