missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Naphthonitrile, TCI America™

$82.50 - $212.36

Chemical Identifiers

CAS 613-46-7
Molecular Formula C11H7N
Molecular Weight (g/mol) 153.184
MDL Number MFCD00016807
InChI Key AZKDTTQQTKDXLH-UHFFFAOYSA-N
Synonym 2-naphthonitrile, 2-cyanonaphthalene, 2-naphthalenecarbonitrile, 2-cyanonaphthylene, 2-naphthalenenitrile, beta-cyanonaphthalene, beta-naphthonitrile, .beta.-naphthonitrile, naphthalene-2-carbonitrile, 2-naphthylcyanide
PubChem CID 11944
IUPAC Name naphthalene-2-carbonitrile
SMILES C1=CC=C2C=C(C=CC2=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N003925G View Documents TCI America
N003925G
25g Each for $212.36
 
N00395G View Documents TCI America
N00395G
5g Each for $82.50
 
Specifications

Chemical Identifiers

613-46-7
153.184
AZKDTTQQTKDXLH-UHFFFAOYSA-N
11944
C1=CC=C2C=C(C=CC2=C1)C#N
C11H7N
MFCD00016807
2-naphthonitrile, 2-cyanonaphthalene, 2-naphthalenecarbonitrile, 2-cyanonaphthylene, 2-naphthalenenitrile, beta-cyanonaphthalene, beta-naphthonitrile, .beta.-naphthonitrile, naphthalene-2-carbonitrile, 2-naphthylcyanide
naphthalene-2-carbonitrile

Specifications

156°C
67°C
613-46-7
MFCD00016807
2-naphthonitrile, 2-cyanonaphthalene, 2-naphthalenecarbonitrile, 2-cyanonaphthylene, 2-naphthalenenitrile, beta-cyanonaphthalene, beta-naphthonitrile, .beta.-naphthonitrile, naphthalene-2-carbonitrile, 2-naphthylcyanide
C1=CC=C2C=C(C=CC2=C1)C#N
153.184
153.18
≥98.0% (GC)
White-Yellow
25g
C11H7N
3439
AZKDTTQQTKDXLH-UHFFFAOYSA-N
naphthalene-2-carbonitrile
11944
Crystal-Powder at 20°C
2-Naphthonitrile
Safety and Handling

Safety and Handling

RTECSNumber : QL5976350

TSCA : Yes