Please login to your online account to display your discounted pricing

2-Naphthonitrile, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  346310050

 View more versions of this product

Catalog No. AC346310050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 153.18
Physical Form Crystalline Powder
Quantity 5g
Color Off-white to beige
RTECS Number QL5976350
TSCA TSCA
EINECS Number 210-344-5
Melting Point 63° to 68°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Packaging Glass bottle
CAS 613-46-7
Health Hazard 2 GHS P Statement Harmful in contact with skin. Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation.
Chemical Name or Material 2-Naphthonitrile
Name Note 97%
Molecular Formula C11H7N
MDL Number MFCD00016807
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-Naphthonitrile
Name Note 97%
CAS 613-46-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H7N
Formula Weight 153.18
MDL Number MFCD00016807
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Harmful in contact with skin. Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation.

SDS & Safety Documents