Please login to your online account to display your discounted pricing

2-Naphthol (Tech.), 97%, ACROS Organics™


Quantity 2kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  422970020

 View more versions of this product

Catalog No. AC422970020

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Color Slightly brownish
Purity Grade Technical Grade
Physical Form Flakes
Formula Weight 144.17
Quantity 2kg
TSCA TSCA
Melting Point 119° to 124°C
RTECS Number QL2975000
EINECS Number 205-182-7
Beilstein 06, 627
Merck Index 15, 6469
CAS Min % 96.0
CAS Max % 100.0
Synonym 2-Hydroxynaphthalene
Packaging Plastic bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.
Chemical Name or Material 2-Naphthol
Health Hazard 1 Warning
Name Note tech., 97%
Linear Formula C10H7OH
Molecular Formula C10H8O
CAS 135-19-3
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-Naphthol
Name Note tech., 97%
CAS 135-19-3
CAS Min % 96.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H8O
Linear Formula C10H7OH
Formula Weight 144.17
Synonym 2-Hydroxynaphthalene
Accessories

Warning

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.

SDS & Safety Documents