Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Naphthoic hydrazide, 98%

$84.97 - $306.31

Chemical Identifiers

Chemical Name or Material 2-Naphthoic hydrazide
CAS 39627-84-4
Molecular Formula C11H10N2O
Formula Weight 186.21
MDL Number MFCD00016810
Beilstein 1950982
Synonym 2-Naphthoylhydrazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0803803

 
alfa aesar™
L0803803
1g Each for $84.97

AAL0803806

 
alfa aesar™
L0803806
5g Each for $306.31
Description & Specifications

Specifications

Formula Weight 186.21
Chemical Name or Material 2-Naphthoic hydrazide
Assay 98%
Molecular Formula C11H10N2O
MDL Number MFCD00016810
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 1950982
Synonym 2-Naphthoylhydrazine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
CAS 39627-84-4
Boiling Point 302° to 304°C (725mmHg)
Melting Point 145° to 149°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.