missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Naphthalenesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 158441000

 View more versions of this product

Catalog No. AC158441000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Naphthalenesulfonyl chloride
Glass bottle
74°C to 78°C
93-11-8
100.0
C10H7ClO2S
11, 173
Solubility in water: insoluble
C1=CC=C2C=C(C=CC2=C1)S(=O)(=O)Cl
226.68
226.68
97%
Authentic
Yellow to Beige
100g
96.0
97%
C10H7SO2Cl
.beta.-naphthalenesulfochloride
OPECTNGATDYLSS-UHFFFAOYSA-N
naphthalene-2-sulfonyl chloride
7125
Powder

Chemical Identifiers

93-11-8
226.68
.beta.-naphthalenesulfochloride
naphthalene-2-sulfonyl chloride
C10H7ClO2S
OPECTNGATDYLSS-UHFFFAOYSA-N
7125
C1=CC=C2C=C(C=CC2=C1)S(=O)(=O)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-219-9

SDS Documents