Please login to your online account to display your discounted pricing

2-Naphthalenesulfonic acid hydrate, 90%, Technical, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC428690050

 View more versions of this product

Catalog No. AC428690050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Physical Form Powder
Assay ≥88%
Formula Weight 208.24
Quantity 5g
Color Gray
Density 1.44g/mL
CAS Min % 88.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Assay Percent Range 90%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Naphthalenesulfonic acid hydrate
Packaging Glass bottle
Molecular Formula C10H8O3S·xH2O
MDL Number MFCD00149271
CAS 76530-12-6
CAS 120-18-3

Specifications

Chemical Name or Material 2-Naphthalenesulfonic acid hydrate
CAS 76530-12-6
CAS Min % 88.0
CAS Max % 100.0
CAS 120-18-3
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C10H8O3S·xH2O
Formula Weight 208.24
MDL Number MFCD00149271
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents