missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Naphthalenemethanol, TCI America™

$47.36 - $139.03

Chemical Identifiers

CAS 1592-38-7
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004124
InChI Key MFGWMAAZYZSWMY-UHFFFAOYSA-N
Synonym 2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na
PubChem CID 74128
ChEBI CHEBI:27615
IUPAC Name naphthalen-2-ylmethanol
SMILES C1=CC=C2C=C(C=CC2=C1)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N08815G View Documents TCI America
N08815G
5g Each for $47.36
 
N088125G View Documents TCI America
N088125G
25g Each for $139.03
 
Specifications

Chemical Identifiers

1592-38-7
158.2
MFGWMAAZYZSWMY-UHFFFAOYSA-N
74128
naphthalen-2-ylmethanol
C11H10O
MFCD00004124
2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na
CHEBI:27615
C1=CC=C2C=C(C=CC2=C1)CO

Specifications

178°C
80°C
1592-38-7
MFCD00004124
MFGWMAAZYZSWMY-UHFFFAOYSA-N
naphthalen-2-ylmethanol
74128
158.20
≥98.0% (GC)
White-Yellow
5g
C11H10O
2-naphthalenemethanol, 2-naphthylmethanol, 2-hydroxymethylnaphthalene, 2-naphthalene methanol, 2-naphthyl methanol, naphthalen-2-yl-methanol, naphthalen-2-yl methanol, 2-naphthylmethan-1-ol, 2-naphthylcarbinol, 2na
C1=CC=C2C=C(C=CC2=C1)CO
158.2
CHEBI:27615
Crystal-Powder at 20°C
2-Naphthalenemethanol
Safety and Handling

Safety and Handling

TSCA : No