missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Naphthaleneboronic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2415714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Naphthaleneboronic acid | |
∼267°C | |
MFCD00236051 | |
2936449 | |
KPTRDYONBVUWPD-UHFFFAOYSA-N | |
naphthalen-2-ylboronic acid | |
2734375 | |
97% |
32316-92-0 | |
C10H9BO2 | |
25 g | |
2-naphthaleneboronic acid, 2-naphthylboronic acid, naphthalene-2-boronic acid, 2-naphthalene boronic acid, 2-naphthaleneylboronic acid, naphthalenyl-2-boronic acid, 2-naphtahaleneboronic acid, beta-naphthylboronic acid, naphthalen-2-boronic acid, naphthalen-2-yl-boranediol | |
OB(O)C1=CC2=CC=CC=C2C=C1 | |
171.99 | |
171.99 |
Chemical Identifiers
32316-92-0 | |
171.99 | |
KPTRDYONBVUWPD-UHFFFAOYSA-N | |
2734375 |
C10H9BO2 | |
MFCD00236051 | |
2-naphthaleneboronic acid, 2-naphthylboronic acid, naphthalene-2-boronic acid, 2-naphthalene boronic acid, 2-naphthaleneylboronic acid, naphthalenyl-2-boronic acid, 2-naphtahaleneboronic acid, beta-naphthylboronic acid, naphthalen-2-boronic acid, naphthalen-2-yl-boranediol | |
OB(O)C1=CC2=CC=CC=C2C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only