Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Propylthiophene, 97%

$77.15 - $77.15

Chemical Identifiers

Chemical Name or Material 2-n-Propylthiophene
CAS 1551-27-5
Molecular Formula C7H10S
Formula Weight 126.22
MDL Number MFCD00014533
UN Number UN1993
Beilstein 105753
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0305206

  SDS
alfa aesar™
L0305206
5g Each for $77.15

AAL0305214

  SDS
alfa aesar™
L0305214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.22
Chemical Name or Material 2-n-Propylthiophene
Assay 97%
Molecular Formula C7H10S
MDL Number MFCD00014533
CAS 1551-27-5
UN Number UN1993
Beilstein 105753
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.
Flash Point 32°C (90°F)
Odor Characteristic
Boiling Point 158° to 160°C
Quantity 5g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS H Statement
H226
Flammable liquid and vapor.