Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(n-Propylthio)nicotinic acid, 97%

$389.43 - $1,245.28

Chemical Identifiers

Chemical Name or Material 2-(n-Propylthio)nicotinic acid
CAS 175135-22-5
Molecular Formula C9H11NO2S
Formula Weight 197.26
MDL Number MFCD00051682
Synonym 2-(n-Propylmercapto)nicotinic acid; 2-(n-Propylthio)pyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1462314

 
alfa aesar™
A1462314
25g Each for $389.43

AAA1462322

 
alfa aesar™
A1462322
100g Each for $1,245.28
Description & Specifications

Specifications

Formula Weight 197.26
Chemical Name or Material 2-(n-Propylthio)nicotinic acid
Assay 97%
Molecular Formula C9H11NO2S
MDL Number MFCD00051682
CAS 175135-22-5
Synonym 2-(n-Propylmercapto)nicotinic acid; 2-(n-Propylthio)pyridine-3-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 159° to 163°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.