missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(n-Propylthio)nicotinic acid, 97%

Manufacturer:  Alfa Aesar™ A1462306

 View more versions of this product

Catalog No. AAA1462306



Specifications

Specifications

2-(n-Propylthio)nicotinic acid
5g
C9H11NO2S
2-propylthio nicotinic acid, 2-n-propylthio nicotinic acid, 2-propylsulfanyl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 2-propylthio, 2-propylthiopyridine-3-carboxylic acid, maybridge1_001579, 2-prop-1-ylsulphanyl pyridine-3-carboxylic acid, 2-propylsulfanyl-nicotinic acid
CCCSC1=C(C=CC=N1)C(=O)O
197.252
197.26
159°C to 163°C
175135-22-5
MFCD00051682
CSMDLVRDBYKTAH-UHFFFAOYSA-N
2-propylsulfanylpyridine-3-carboxylic acid
2774526
97%

Chemical Identifiers

175135-22-5
197.252
CSMDLVRDBYKTAH-UHFFFAOYSA-N
2774526
CCCSC1=C(C=CC=N1)C(=O)O
C9H11NO2S
MFCD00051682
2-propylthio nicotinic acid, 2-n-propylthio nicotinic acid, 2-propylsulfanyl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 2-propylthio, 2-propylthiopyridine-3-carboxylic acid, maybridge1_001579, 2-prop-1-ylsulphanyl pyridine-3-carboxylic acid, 2-propylsulfanyl-nicotinic acid
2-propylsulfanylpyridine-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents