Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(n-Propylthio)nicotinamide, 97%

$241.08 - $241.08

Chemical Identifiers

Chemical Name or Material 2-(n-Propylthio)nicotinamide
CAS 175135-26-9
Molecular Formula C9H12N2OS
Formula Weight 196.27
MDL Number MFCD00052904
Synonym 2-(n-Propylthio)pyridine-3-carboxamide; 2-(n-Propylmercapto)nicotinamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2525903

  SDS
alfa aesar™
B2525903
1g Each for $241.08
Description & Specifications

Specifications

Formula Weight 196.27
Chemical Name or Material 2-(n-Propylthio)nicotinamide
Molecular Formula C9H12N2OS
MDL Number MFCD00052904
Quantity 1g
Assay 97%
Synonym 2-(n-Propylthio)pyridine-3-carboxamide; 2-(n-Propylmercapto)nicotinamide
Melting Point 146° to 148°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 175135-26-9
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.