Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(n-Propylthio)ethanol, 98%

Chemical Identifiers

Chemical Name or Material 2-(n-Propylthio)ethanol
CAS 22812-90-4
Molecular Formula C5H12OS
Linear Formula CH3(CH2)2S(CH2)2OH
Formula Weight 120.21
MDL Number MFCD00014041
UN Number UN1760
Beilstein 1731984
Synonym 2-Hydroxyethyl n-propyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1252806

  SDS
alfa aesar™
L1252806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1252814

  SDS
alfa aesar™
L1252814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.21
Molecular Formula C5H12OS
Chemical Name or Material 2-(n-Propylthio)ethanol
CAS 22812-90-4
Synonym 2-Hydroxyethyl n-propyl sulfide
Boiling Point 199° to 200°C
Beilstein 1731984
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
Assay 98%
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
UN Number UN1760
MDL Number MFCD00014041
Quantity 5g
Odor Unpleasant
Flash Point 98°C (208°F)
Linear Formula CH3(CH2)2S(CH2)2OH
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.