missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(n-Propylthio)ethanol, 98%

Manufacturer:  Alfa Aesar™ L1252814

 View more versions of this product

Catalog No. AAL1252814



Specifications

Specifications

2-(n-Propylthio)ethanol
Unpleasant
25g
C5H12OS
MFCD00014041
1731984
KCWWXXYQPUDKBX-UHFFFAOYSA-N
2-propylsulfanylethanol
89850
98%
98°C (208°F)
199°C to 200°C
22812-90-4
CH3(CH2)2S(CH2)2OH
UN1760
2-propylthio ethanol, 2-hydroxyethyl propyl sulfide, 2-n-propylthio ethanol, 2-hydroxyethyl n-propyl sulfide, 2-propylsulfanyl ethanol, 2-propylthioethan-1-ol, acmc-209fyz, ethanol, 2-propylthio, 2-propylsulfanyl ethanol #, 2-hydroxyethyln-propylsulfide
CCCSCCO
120.21
120.21

Chemical Identifiers

22812-90-4
120.21
KCWWXXYQPUDKBX-UHFFFAOYSA-N
89850
CCCSCCO
C5H12OS
MFCD00014041
2-propylthio ethanol, 2-hydroxyethyl propyl sulfide, 2-n-propylthio ethanol, 2-hydroxyethyl n-propyl sulfide, 2-propylsulfanyl ethanol, 2-propylthioethan-1-ol, acmc-209fyz, ethanol, 2-propylthio, 2-propylsulfanyl ethanol #, 2-hydroxyethyln-propylsulfide
2-propylsulfanylethanol
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P261-P280-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.

Documents