missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Propylthiazole, 99%

Manufacturer:  Alfa Aesar™ A1157406

 View more versions of this product

Catalog No. AAA1157406



Specifications

Specifications

2-n-Propylthiazole
62°C (15mmHg)
17626-75-4
MFCD00053154
2-propylthiazole, thiazole, 2-propyl, 2-n-propylthiazole, unii-t395979vrl, 2-propylthiazol, thiazole,2-propyl, 2-normal-propyl thiazole, acmc-209eb6, #
CCCC1=NC=CS1
127.205
127.2
54°C (129°F)
5g
C6H9NS
UN1993
CMOIEFFAOUQJPS-UHFFFAOYSA-N
2-propyl-1,3-thiazole
87198
99%

Chemical Identifiers

17626-75-4
127.205
CMOIEFFAOUQJPS-UHFFFAOYSA-N
87198
CCCC1=NC=CS1
C6H9NS
MFCD00053154
2-propylthiazole, thiazole, 2-propyl, 2-n-propylthiazole, unii-t395979vrl, 2-propylthiazol, thiazole,2-propyl, 2-normal-propyl thiazole, acmc-209eb6, #
2-propyl-1,3-thiazole
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

SDS Documents