Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Propylthiazole, 99%

Chemical Identifiers

Chemical Name or Material 2-n-Propylthiazole
CAS 17626-75-4
Molecular Formula C6H9NS
Formula Weight 127.2
MDL Number MFCD00053154
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1157406

  SDS
alfa aesar™
A1157406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.2
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 54°C (129°F)
CAS 17626-75-4
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
UN Number UN1993
Assay 99%
Quantity 5g
Boiling Point 62°C (15mmHg)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Chemical Name or Material 2-n-Propylthiazole
Molecular Formula C6H9NS
Health Hazard 1 Warning
TSCA TSCA
MDL Number MFCD00053154
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.