Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Propylresorcinol, 98%

$115.12 - $337.51

Chemical Identifiers

Chemical Name or Material 2-n-Propylresorcinol
CAS 13331-19-6
Molecular Formula C9H12O2
Formula Weight 152.19
MDL Number MFCD00100568
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2702403

  SDS
alfa aesar™
H2702403
1g Each for $115.12

AAH2702406

  SDS
alfa aesar™
H2702406
5g Each for $337.51
Description & Specifications

Specifications

Formula Weight 152.19
Chemical Name or Material 2-n-Propylresorcinol
Assay 98%
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
MDL Number MFCD00100568
Odor Phenol-like
Flash Point 135°C (275°F)
Molecular Formula C9H12O2
Boiling Point 280°C
Melting Point 101° to 105°C
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
CAS 13331-19-6

Applications
2-n-propylresorcinol is a pharmaceutical intermediate.

Solubility
Solightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.