missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Propylphenol, 97%

Manufacturer:  Alfa Aesar™ A1108806

 View more versions of this product

Catalog No. AAA1108806



Specifications

Specifications

2-n-Propylphenol
Phenol-like
5g
C9H12O
UN3145
2-n-propylphenol, o-propylphenol, phenol, 2-propyl, phenol, o-propyl, 1-hydroxy-2-n-propylbenzene, 1-2-hydroxyphenyl propane, propylphenol, 2-propyl-phenol, o-n-propylphenol, unii-333r6f6t3n
CCCC1=CC=CC=C1O
136.194
136.19
93°C (199°F)
214°C
644-35-9
MFCD00002251
1363932
LCHYEKKJCUJAKN-UHFFFAOYSA-N
2-propylphenol
12570
97%

Chemical Identifiers

644-35-9
136.194
LCHYEKKJCUJAKN-UHFFFAOYSA-N
12570
CCCC1=CC=CC=C1O
C9H12O
MFCD00002251
2-n-propylphenol, o-propylphenol, phenol, 2-propyl, phenol, o-propyl, 1-hydroxy-2-n-propylbenzene, 1-2-hydroxyphenyl propane, propylphenol, 2-propyl-phenol, o-n-propylphenol, unii-333r6f6t3n
2-propylphenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.

RTECSNumber : SM8600000

TSCA : TSCA

SDS Documents