Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Propylphenol, 97%

Chemical Identifiers

Chemical Name or Material 2-n-Propylphenol
CAS 644-35-9
Molecular Formula C9H12O
Formula Weight 136.19
MDL Number MFCD00002251
UN Number UN3145
Beilstein 1363932
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1108814

  SDS
alfa aesar™
A1108814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.19
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Phenol-like
RTECS Number SM8600000
Chemical Name or Material 2-n-Propylphenol
Assay 97%
Quantity 25g
CAS 644-35-9
Boiling Point 214°C
Health Hazard 1 Danger
Beilstein 1363932
Health Hazard 2 GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.
Flash Point 93°C (199°F)
MDL Number MFCD00002251
Molecular Formula C9H12O
UN Number UN3145
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.