Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Propylfuran, 99%

Chemical Identifiers

Chemical Name or Material 2-n-Propylfuran
CAS 4229-91-8
Molecular Formula C6H8O
Formula Weight 110.16
MDL Number MFCD00047070
UN Number UN3271
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2170306

  SDS
alfa aesar™
B2170306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2170314

  SDS
alfa aesar™
B2170314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 110.16
Chemical Name or Material 2-n-Propylfuran
Assay 99%
Molecular Formula C6H8O
MDL Number MFCD00047070
CAS 4229-91-8
UN Number UN3271
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
Flash Point 24°C (75°F)
Boiling Point 110° to 111°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.

2° to 8°C