Learn More
2-n-Propyl-1-heptanol, 98%, Thermo Scientific Chemicals
$378.42 - $1660.21
Chemical Identifiers
CAS | 10042-59-8 |
---|---|
Molecular Formula | C10H22O |
Molecular Weight (g/mol) | 158.285 |
MDL Number | MFCD00046768 |
InChI Key | YLQLIQIAXYRMDL-UHFFFAOYSA-N |
Synonym | 2-propyl-1-heptanol, 1-heptanol, 2-propyl, 2-propylheptanol, 2-n-propyl-1-heptanol, 2-propylheptyl alcohol, 2-propyl heptanol, 2-propyl-heptan-1-ol, acmc-20ann4, 2-propyl-1-heptanol #, dsstox_cid_9302 |
PubChem CID | 24847 |
IUPAC Name | 2-propylheptan-1-ol |
SMILES | CCCCCC(CCC)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5034003
|
Thermo Scientific Chemicals
H5034003 |
1 g |
Each for $378.42
|
|
|||||
AAH5034006
|
Thermo Scientific Chemicals
H5034006 |
5 g |
Each for $1,660.21
|
|
|||||
Description
2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.
Solubility
Insoluble in water. Miscible with most common organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
10042-59-8 | |
158.285 | |
YLQLIQIAXYRMDL-UHFFFAOYSA-N | |
24847 | |
CCCCCC(CCC)CO |
C10H22O | |
MFCD00046768 | |
2-propyl-1-heptanol, 1-heptanol, 2-propyl, 2-propylheptanol, 2-n-propyl-1-heptanol, 2-propylheptyl alcohol, 2-propyl heptanol, 2-propyl-heptan-1-ol, acmc-20ann4, 2-propyl-1-heptanol #, dsstox_cid_9302 | |
2-propylheptan-1-ol |
Specifications
10042-59-8 | |
MFCD00046768 | |
2-propyl-1-heptanol, 1-heptanol, 2-propyl, 2-propylheptanol, 2-n-propyl-1-heptanol, 2-propylheptyl alcohol, 2-propyl heptanol, 2-propyl-heptan-1-ol, acmc-20ann4, 2-propyl-1-heptanol #, dsstox_cid_9302 | |
YLQLIQIAXYRMDL-UHFFFAOYSA-N | |
2-propylheptan-1-ol | |
24847 | |
98% |
C10H22O | |
1 g | |
Insoluble in water. Miscible with most common organic solvents. | |
CCCCCC(CCC)CO | |
158.285 | |
158.28 | |
2-n-Propyl-1-heptanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 233-126-1
RTECSNumber : MJ4800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only