Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Propoxybenzoic acid, 98+%

$44.15 - $640.38

Chemical Identifiers

Chemical Name or Material 2-n-Propoxybenzoic acid
CAS 2100-31-4
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD01075687
Beilstein 3089998
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2458103

  SDS
alfa aesar™
B2458103
1g Each for $44.15

AAB2458114

  SDS
alfa aesar™
B2458114
25g Each for $640.38
Description & Specifications

Specifications

Formula Weight 180.2
Melting Point 36° to 40°C
Health Hazard 1 Warning
Chemical Name or Material 2-n-Propoxybenzoic acid
Beilstein 3089998
Assay ≥98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
MDL Number MFCD01075687
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H12O3
CAS 2100-31-4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.