Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Propoxybenzaldehyde, 97+%

$69.92 - $250.12

Chemical Identifiers

Chemical Name or Material 2-n-Propoxybenzaldehyde
CAS 7091-12-5
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00046150
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5047303

  SDS
alfa aesar™
H5047303
1g Each for $250.12

AAH50473MD

  SDS
alfa aesar™
H50473MD
250mg Each for $69.92
Description & Specifications

Specifications

Formula Weight 164.2
Quantity 1g
Molecular Formula C10H12O2
Assay ≥97%
Health Hazard 1 Warning
Chemical Name or Material 2-n-Propoxybenzaldehyde
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 7091-12-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00046150
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.