Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Pentylthiophene, 98%

$65.09 - $65.09

Chemical Identifiers

Chemical Name or Material 2-n-Pentylthiophene
CAS 4861-58-9
Molecular Formula C9H14S
Formula Weight 154.28
MDL Number MFCD00041017
Beilstein 107941
Synonym 2-n-Amylthiophene; 1-(2-Thienyl)pentane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2188506

  SDS
alfa aesar™
B2188506
5g Each for $65.09
Description & Specifications

Specifications

Formula Weight 154.28
Chemical Name or Material 2-n-Pentylthiophene
TSCA TSCA
Assay 98%
Molecular Formula C9H14S
MDL Number MFCD00041017
CAS 4861-58-9
Beilstein 107941
Synonym 2-n-Amylthiophene; 1-(2-Thienyl)pentane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 75°C (167°F)
Boiling Point 198°C
Quantity 5g

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also used as flavoring agent for food.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.