Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Pentylpyrazine, 97%


Quantity 1g

Manufacturer: alfa aesar™  L0303903

 View more versions of this product

Catalog No. AAL0303903

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 150.23
Assay 97%
Molecular Formula C9H14N2
CAS 6303-75-9
Health Hazard 1 Warning
Chemical Name or Material 2-n-Pentylpyrazine
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Synonym n-Amylpyrazine; 1-Pyrazinylpentane
Boiling Point 218°C
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Quantity 1g
Beilstein 508952
Flash Point 81°C (178°F)
MDL Number MFCD00014614

Specifications

Chemical Name or Material 2-n-Pentylpyrazine
CAS 6303-75-9
Molecular Formula C9H14N2
Formula Weight 150.23
MDL Number MFCD00014614
Beilstein 508952
Synonym n-Amylpyrazine; 1-Pyrazinylpentane
Accessories

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

SDS & Safety Documents