Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Pentylpyrazine, 97%

$103.76 - $357.11

Chemical Identifiers

Chemical Name or Material 2-n-Pentylpyrazine
CAS 6303-75-9
Molecular Formula C9H14N2
Formula Weight 150.23
MDL Number MFCD00014614
Beilstein 508952
Synonym n-Amylpyrazine; 1-Pyrazinylpentane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0303903

 
alfa aesar™
L0303903
1g Each for $103.76

AAL0303906

 
alfa aesar™
L0303906
5g Each for $357.11
Description & Specifications

Specifications

Formula Weight 150.23
Chemical Name or Material 2-n-Pentylpyrazine
Assay 97%
Molecular Formula C9H14N2
CAS 6303-75-9
Health Hazard 1 Warning
Synonym n-Amylpyrazine; 1-Pyrazinylpentane
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Boiling Point 218°C
MDL Number MFCD00014614
Beilstein 508952
Flash Point 81°C (178°F)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.