Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-n-Octylthiophene, 98%

$101.57 - $601.20

Chemical Identifiers

Chemical Name or Material 2-n-Octylthiophene
CAS 880-36-4
Molecular Formula C12H20S
Formula Weight 196.36
MDL Number MFCD00041015
Beilstein 114488
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1367206

  SDS
alfa aesar™
A1367206
5g Each for $101.57

AAA1367214

  SDS
alfa aesar™
A1367214
25g Each for $336.01

AAA1367218

  SDS
alfa aesar™
A1367218
50g Each for $601.20
Description & Specifications

Specifications

Formula Weight 196.36
Quantity 5g
Molecular Formula C12H20S
Chemical Name or Material 2-n-Octylthiophene
Assay 98%
Flash Point >100°C (212°F)
Beilstein 114488
CAS 880-36-4
MDL Number MFCD00041015
Boiling Point 127° to 129°C (12mmHg)