missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-n-Octyl-1-dodecanol 90.0+%, TCI America™

Manufacturer:  TCI America O042925ML

 View more versions of this product

Catalog No. O042925ML



Specifications

Specifications

2-n-Octyl-1-dodecanol
Colorless
25mL
C20H42O
LEACJMVNYZDSKR-UHFFFAOYSA-N
2-octyldodecan-1-ol
21414
Clear Liquid at 20°C
161°C
1°C
5333-42-6
MFCD01314208
CCCCCCCCCCC(CCCCCCCC)CO
298.555
298.56
≥90.0% (GC)

Chemical Identifiers

5333-42-6
298.555
LEACJMVNYZDSKR-UHFFFAOYSA-N
2-octyldodecan-1-ol
C20H42O
MFCD01314208
21414
CCCCCCCCCCC(CCCCCCCC)CO
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

RTECSNumber : JR4240000

TSCA : Yes

Documents